2,6-Dibromoanisole ≥97%

Supplier: AOBChem USA

Warning

16449-25G 16449-5G
76896-462EA 268.41 USD
76896-462 76896-464
2,6-Dibromoanisole ≥97%
2,6-Dibromoanisole

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
Storage Temperature: Ambient
MDL Number: MFCD00093270
CAS Number: 38603-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR