2-Fluoro-5-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

16405-1G 16405-25G 16405-500MG 16405-5G
76896-436EA 409.51 USD
76896-436 76896-438 76896-440 76896-442
2-Fluoro-5-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
2-Fluoro-5-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula: C₁₂H₁₇BFNO₂
MW: 237.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08063096
CAS Number: 755027-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR