2-Chloro-6-fluoro-3-(methoxymethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16390-1G 16390-25G 16390-5G
76896-412EA 64.72 USD
76896-412 76896-414 76896-416
2-Chloro-6-fluoro-3-(methoxymethoxy)phenylboronic acid ≥97%
2-Chloro-6-fluoro-3-(methoxymethoxy)phenylboronic acid
Formula: C₈H₉BClFO₄
MW: 234.417 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617262
CAS Number: 1451392-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR