2,6-Dichloro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16326-1G 16326-25G 16326-500MG 16326-5G
76896-390EA 142.17 USD
76896-390 76896-392 76896-394 76896-396
2,6-Dichloro-3-methylbenzaldehyde ≥97%
2,6-Dichloro-3-methylbenzaldehyde
Formula: C₈H₆Cl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09999657
CAS Number: 60611-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR