2,6-Dichloro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16315-25G 16315-5G
76896-380EA 453.01 USD
76896-380 76896-382
2,6-Dichloro-4-iodobenzaldehyde ≥97%
2,6-Dichloro-4-iodobenzaldehyde
Formula: C₇H₃Cl₂IO
MW: 300.909 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27946243
CAS Number: 177167-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR