2-Bromo-6-fluoro-3-methoxybenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
16297-1G
16297-25G
16297-5G
76896-370EA
114.58
USD
76896-370
76896-372
76896-374
2-Bromo-6-fluoro-3-methoxybenzaldehyde ≥97%
2-Bromo-6-fluoro-3-methoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11856027 CAS Number: 154650-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...