2-(Dimethylamino)pyrimidine-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Dimethylamino-pyrimidine-5-carbaldehyde

16114-1G 16114-25G 16114-5G
76896-338EA 76.39 USD
76896-338 76896-340 76896-342
2-(Dimethylamino)pyrimidine-5-carbaldehyde ≥97%
2-(Dimethylamino)pyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₉N₃O
MW: 151.166 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03446733
CAS Number: 55551-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR