2-Bromo-3,5-bis(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

16091-1G 16091-25G 16091-500MG 16091-5G
76896-328EA 173.99 USD
76896-328 76896-330 76896-332 76896-334
2-Bromo-3,5-bis(trifluoromethyl)benzoic acid ≥97%
2-Bromo-3,5-bis(trifluoromethyl)benzoic acid
Formula: C₉H₃BrF₆O₂
MW: 337.013 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181635
CAS Number: 505084-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR