2,6-Difluoro-4-carboxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Borono-3,5-difluorobenzoic acid

16041-1G 16041-25G 16041-500MG 16041-5G
76896-314EA 183.54 USD
76896-314 76896-316 76896-318 76896-320
2,6-Difluoro-4-carboxyphenylboronic acid ≥97%
2,6-Difluoro-4-carboxyphenylboronic acid
Formula: C₇H₅BF₂O₄
MW: 201.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17214253
CAS Number: 1031857-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR