2-Chloro-6-fluoro-5-hydroxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

15991-1G 15991-25G 15991-500MG 15991-5G
76896-290EA 193.09 USD
76896-290 76896-292 76896-294 76896-296
2-Chloro-6-fluoro-5-hydroxyphenylboronic acid ≥95%
2-Chloro-6-fluoro-5-hydroxyphenylboronic acid
Formula: C₆H₅BClFO₃
MW: 190.365 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105351
CAS Number: 958646-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR