2-(Ethoxycarbonyl)-5-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Ethoxycabonyl-5-fluorophenylboronic acid, (2-(Ethoxycarbonyl)-5-fluorophenyl)boronic acid

15915-1G 15915-25G 15915-5G
76896-256EA 99.73 USD
76896-256 76896-258 76896-260
2-(Ethoxycarbonyl)-5-fluorophenylboronic acid ≥97%
2-(Ethoxycarbonyl)-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
Storage Temperature: Ambient
MDL Number: MFCD09878342
CAS Number: 957062-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR