2-Chloro-3,6-difluorophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

15893-1G 15893-25G 15893-500MG 15893-5G
76896-244EA 225.98 USD
76896-244 76896-246 76896-248 76896-250
2-Chloro-3,6-difluorophenylboronic acid ≥97% research grade
2-Chloro-3,6-difluorophenylboronic acid
Formula: C₆H₄BClF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18415644
CAS Number: 2377608-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR