2-Methoxy-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-5-trifluoromethylphenol

15879-1G 15879-25G 15879-5G
76896-236EA 106.09 USD
76896-236 76896-238 76896-240
2-Methoxy-5-(trifluoromethyl)phenol ≥97%
2-Methoxy-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₈H₇F₃O₂
MW: 192.135 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09832299
CAS Number: 349-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR