2-Fluoro-6-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-2-hydroxybenzotrifluoride

15847-25G 15847-5G
76896-226EA 371.32 USD
76896-226 76896-228
2-Fluoro-6-(trifluoromethyl)phenol ≥97%
2-Fluoro-6-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236300
CAS Number: 239135-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR