2-Chloro-6-(trifluoromethyl)pyridine-3-boronic Acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloro-6-(trifluoromethyl)pyridin-3-yl)boronic acid, 2-Chloro-6-trifluoromethylpyridine-3-boronic acid

15831-1G 15831-25G 15831-500MG 15831-5G
76896-216EA 173.99 USD
76896-216 76896-218 76896-220 76896-222
2-Chloro-6-(trifluoromethyl)pyridine-3-boronic Acid ≥97%
2-Chloro-6-(trifluoromethyl)pyridine-3-boronic Acid
Formula: C₆H₄BClF₃NO₂
MW: 225.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696211
CAS Number: 205240-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR