2-Chloro-6-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15807-25G 15807-5G
76896-204EA 289.63 USD
76896-204 76896-206
2-Chloro-6-methoxybenzaldehyde ≥97%
2-Chloro-6-methoxybenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.5900 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059233
CAS Number: 29866-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR