2-Chloro-4-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloro-4-(trifluoromethoxy)phenyl)boronic acid, 2-Chloro-4-trifluoromethoxyphenylboronic acid

15763-1G 15763-25G 15763-500MG 15763-5G
76896-170EA 245.07 USD
76896-170 76896-172 76896-174 76896-176
2-Chloro-4-(trifluoromethoxy)phenylboronic acid ≥97%
2-Chloro-4-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₃
MW: 240.3720 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11040272
CAS Number: 345226-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR