2-Bromo-6-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15744-1G 15744-25G 15744-5G
76896-148EA 62.6 USD
76896-148 76896-150 76896-152
2-Bromo-6-(trifluoromethoxy)benzaldehyde ≥97%
2-Bromo-6-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11856022
CAS Number: 1114809-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR