2-Chloro-6-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-6-fluoro-3-tolylaldehyde, 2-Chloro-6-fluoro-m-tolylaldehyde, 2-Chloro-6-fluoro-m-tolualdehyde, 2-Chloro-6-fluoro-3-tolualdehyde

15641-25G 15641-5G
76896-116EA 225.98 USD
76896-116 76896-118
2-Chloro-6-fluoro-3-methylbenzaldehyde ≥97%
2-Chloro-6-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MW: 172.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631360
CAS Number: 104451-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR