2-Chloro-N-methoxy-N-methylisonicotinamide ≥97%

Supplier: AOBChem USA

Warning

154867-1G 154867-25G 154867-500MG 154867-5G
76895-992EA 124.13 USD
76895-992 76895-994 76895-996 76895-998
2-Chloro-N-methoxy-N-methylisonicotinamide ≥97%
2-Chloro-N-methoxy-N-methylisonicotinamide
Formula: C₈H₉ClN₂O₂
MW: 200.6222 g/mol
Storage Temperature: Ambient
CAS Number: 250263-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR