(2,6-Difluoro-4-formylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoro-4-formylphenylboronic acid

15465-25G 15465-5G
76895-984EA 488.02 USD
76895-984 76895-986
(2,6-Difluoro-4-formylphenyl)boronic acid ≥97% research grade
(2,6-Difluoro-4-formylphenyl)boronic acid
Formula: C₇H₅BF₂O₃
MW: 185.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697422
CAS Number: 871125-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR