2-Bromo-3-chloro-6-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

15455-1G 15455-25G 15455-500MG 15455-5G
76895-970EA 393.6 USD
76895-970 76895-972 76895-974 76895-976
2-Bromo-3-chloro-6-fluorobenzoic acid ≥97%
2-Bromo-3-chloro-6-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28141929
CAS Number: 1805575-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR