2-Bromo-1-isopropoxy-4-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

15453-1G 15453-25G 15453-500MG 15453-5G
76895-956EA 350.1 USD
76895-956 76895-958 76895-960 76895-962
2-Bromo-1-isopropoxy-4-(trifluoromethoxy)benzene ≥97%
2-Bromo-1-isopropoxy-4-(trifluoromethoxy)benzene
Formula: C₁₀H₁₀BrF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08059526
CAS Number: 200956-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR