2,6-Dibromo-4-(trifluoromethoxy)phenol ≥97%

Supplier: AOBChem USA

Warning

15444-1G 15444-25G 15444-500MG 15444-5G
76895-928EA 159.14 USD
76895-928 76895-930 76895-932 76895-934
2,6-Dibromo-4-(trifluoromethoxy)phenol ≥97%
2,6-Dibromo-4-(trifluoromethoxy)phenol
Formula: C₇H₃Br₂F₃O₂
MW: 335.901 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059524
CAS Number: 914635-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR