2-Chloro-6-fluoro-3-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15419-25G 15419-5G
76895-920EA 481.65 USD
76895-920 76895-922
2-Chloro-6-fluoro-3-hydroxyphenylboronic acid ≥97%
2-Chloro-6-fluoro-3-hydroxyphenylboronic acid
Formula: C₆H₅BClFO₃
MW: 190.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800886
CAS Number: 957121-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR