2-Trifluoromethyl-5-chlorophenol ≥97%

Supplier: AOBChem USA

Warning

15357-1G 15357-25G 15357-5G 15357-500MG
76895-910EA 84.88 USD
76895-910 76895-912 76895-914 77648-744
2-Trifluoromethyl-5-chlorophenol ≥97%
2-Trifluoromethyl-5-chlorophenol
Formula: C₇H₄ClF₃O
MW: 196.554 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520659
CAS Number: 106877-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR