2-Bromo-1-chloro-5-fluoro-3-methylbenzene 97%

Supplier: AOBChem USA

Warning

15328-1G 15328-25G 15328-5G 15328-500MG
76895-888EA 106.09 USD
76895-888 76895-890 76895-892 77644-666
2-Bromo-1-chloro-5-fluoro-3-methylbenzene 97%
2-Bromo-1-chloro-5-fluoro-3-methylbenzene
Formula: C₇H₅BrClF
Storage Temperature: Refrigerator
MDL Number: MFCD16817668
CAS Number: 1242339-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR