2-Bromo-6-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15315-25G 15315-5G
76895-864EA 559.1 USD
76895-864 76895-866
2-Bromo-6-fluoro-4-methylbenzaldehyde ≥97%
2-Bromo-6-fluoro-4-methylbenzaldehyde
Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD22056736
CAS Number: 1370025-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR