2-Fluoro-5-(trifluoromethyl)anisole ≥97%

Supplier: AOBChem USA

Warning

15301-25G 15301-5G
76895-856EA 215.37 USD
76895-856 76895-858
2-Fluoro-5-(trifluoromethyl)anisole ≥97%
2-Fluoro-5-(trifluoromethyl)anisole

Precaución: for Research Use Only

Formula: C₈H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD01631564
CAS Number: 261951-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR