2-Methyl-3-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methyl-3-trifluoromethyl-phenylboronic acid

15265-1G 15265-25G 15265-500MG 15265-5G
76895-844EA 126.25 USD
76895-844 76895-846 76895-848 76895-850
2-Methyl-3-(trifluoromethyl)phenylboronic acid ≥97%
2-Methyl-3-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₂
MW: 203.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042430
CAS Number: 947533-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR