2,6-Dichloro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15243-1G 15243-25G 15243-5G
76895-826EA 77.45 USD
76895-826 76895-828 76895-830
2,6-Dichloro-4-methylbenzaldehyde ≥97%
2,6-Dichloro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD09753749
CAS Number: 116070-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR