2,5-Dichloro-4-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15239-25G 15239-5G
76895-816EA 434.97 USD
76895-816 76895-818
2,5-Dichloro-4-fluorobenzaldehyde ≥97%
2,5-Dichloro-4-fluorobenzaldehyde
Formula: C₇H₃Cl₂FO
MW: 193.003 g/mol
Storage Temperature: Ambient
CAS Number: 230642-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR