2-Bromo-6-chloro-4-fluoroanisole ≥97%

Supplier: AOBChem USA

Warning

15225-25G 15225-5G
76895-808EA 268.41 USD
76895-808 76895-810
2-Bromo-6-chloro-4-fluoroanisole ≥97%
2-Bromo-6-chloro-4-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115952
CAS Number: 222712-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR