(2,6-Dichloro-4-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dichloro-4-fluorophenylboronic acid

15206-1G 15206-25G 15206-500MG 15206-5G
76895-790EA 201.58 USD
76895-790 76895-792 76895-794 76895-796
(2,6-Dichloro-4-fluorophenyl)boronic acid ≥97%
(2,6-Dichloro-4-fluorophenyl)boronic acid
Formula: C₆H₄BCl₂FO₂
MW: 208.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526365
CAS Number: 1451392-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR