2-Bromo-3-trifluoromethylphenylboronic acid ≥96%

Supplier: AOBChem USA

Warning

15183-1G 15183-25G 15183-5G
76895-742EA 96.55 USD
76895-742 76895-744 76895-746
2-Bromo-3-trifluoromethylphenylboronic acid ≥96%
2-Bromo-3-trifluoromethylphenylboronic acid
Formula: C₇H₅BBrF₃O₂
MW: 268.824 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11111902
CAS Number: 1451393-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR