2-Bromo-5-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15159-25G 15159-5G
76895-708EA 321.46 USD
76895-708 76895-710
2-Bromo-5-methoxyphenylboronic acid ≥97%
2-Bromo-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₃
Storage Temperature: Ambient
MDL Number: MFCD06659859
CAS Number: 89694-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR