2-Bromo-5-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

15155-1G 15155-25G 15155-5G
76895-692EA 62.6 USD
76895-692 76895-694 76895-696
2-Bromo-5-fluorophenylboronic acid ≥95%
2-Bromo-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
Storage Temperature: Ambient
MDL Number: MFCD08690232
CAS Number: 959996-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR