2-Hydroxymethyl-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15075-1G 15075-25G 15075-500MG 15075-5G
76895-644EA 340.55 USD
76895-644 76895-646 76895-648 76895-650
2-Hydroxymethyl-4-methylphenylboronic acid ≥97%
2-Hydroxymethyl-4-methylphenylboronic acid
Formula: C₈H₁₁BO₃
MW: 165.982 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22628186
CAS Number: 1451391-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR