2-Bromo-5-fluoro-4-methylaniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-5-fluoro-p-toluidine

15046-25G 15046-5G
76895-638EA 279.02 USD
76895-638 76895-640
2-Bromo-5-fluoro-4-methylaniline ≥97%
2-Bromo-5-fluoro-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MW: 204.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00142871
CAS Number: 202865-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR