2-Bromo-1,3-difluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

149250-1G 149250-25G 149250-500MG 149250-5G
76895-626EA 338.43 USD
76895-626 76895-628 76895-630 76895-632
2-Bromo-1,3-difluoro-4-methylbenzene ≥97%
2-Bromo-1,3-difluoro-4-methylbenzene
Formula: C₇H₅BrF₂
MW: 207.0154 g/mol
Storage Temperature: Ambient
CAS Number: 847502-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR