2-Carboxythiophene-5-boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

14701-25G 14701-5G
76895-590EA 240.83 USD
76895-590 76895-592
2-Carboxythiophene-5-boronic acid ≥95% research grade
2-Carboxythiophene-5-boronic acid
Formula: C₅H₅BO₄S
Storage Temperature: Ambient
MDL Number: MFCD02323211
CAS Number: 465515-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR