2-Bromo-4-methyl-1-(propan-2-yloxy)benzene ≥97%

Supplier: AOBChem USA

Warning

14564-25G 14564-5G
76895-568EA 453.01 USD
76895-568 76895-570
2-Bromo-4-methyl-1-(propan-2-yloxy)benzene ≥97%
2-Bromo-4-methyl-1-(propan-2-yloxy)benzene
Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11156049
CAS Number: 13178-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR