2-Fluoro-5-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-5-hydroxyphenyl)boronic acid

14536-25G 14536-5G
76895-548EA 552.73 USD
76895-548 76895-550
2-Fluoro-5-hydroxyphenylboronic acid ≥97%
2-Fluoro-5-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₃
MW: 155.919 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11855845
CAS Number: 1150114-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR