2-Hydroxy-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Trifluoromethyl)salicylic acid, 3-Carboxy-2-hydroxybenzotrifluoride

14527-1G 14527-25G 14527-5G
76895-526EA 110.34 USD
76895-526 76895-528 76895-530
2-Hydroxy-3-(trifluoromethyl)benzoic acid ≥97%
2-Hydroxy-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD06203614
CAS Number: 251300-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR