2-Fluoro-3-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

14521-1G 14521-25G 14521-5G
76895-518EA 106.09 USD
76895-518 76895-520 76895-522
2-Fluoro-3-hydroxybenzoic acid ≥97%
2-Fluoro-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅FO₃
MW: 156.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862066
CAS Number: 91658-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR