2-Bromo-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

14490-25G 14490-5G
76895-506EA 190.97 USD
76895-506 76895-508
2-Bromo-5-methylbenzaldehyde ≥95%
2-Bromo-5-methylbenzaldehyde
Formula: C₈H₇BrO
Storage Temperature: Refrigerator
MDL Number: MFCD08669550
CAS Number: 90221-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR