2-Chloro-3-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14482-1G 14482-25G 14482-500MG 14482-5G
76895-494EA 380.87 USD
76895-494 76895-496 76895-498 76895-500
2-Chloro-3-hydroxyphenylboronic acid ≥97%
2-Chloro-3-hydroxyphenylboronic acid
Formula: C₆H₆BClO₃
Storage Temperature: Refrigerator
CAS Number: 854778-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR