(2-Methoxy-3-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-3-methylphenylboronic acid

14426-1G 14426-25G 14426-5G
76895-466EA 117.76 USD
76895-466 76895-468 76895-470
(2-Methoxy-3-methylphenyl)boronic acid ≥97%
(2-Methoxy-3-methylphenyl)boronic acid
Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06801731
CAS Number: 909187-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR