2-Chloro-(5-methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-(methoxycarbonyl)phenylboronic acid, (2-Chloro-5-(methoxycarbonyl)phenyl)boronic acid, 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid

14378-1G 14378-25G 14378-5G
76895-448EA 91.24 USD
76895-448 76895-450 76895-452
2-Chloro-(5-methoxycarbonyl)phenylboronic acid ≥97%
2-Chloro-(5-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BClO₄
MW: 214.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689528
CAS Number: 913835-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR