2,6-Dichloro-4-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,6-Dichloropyridin-4-yl)boronic acid, 2,6-Dichloropyridine-4-boronic acid

14345-25G 14345-5G
76895-440EA 453.01 USD
76895-440 76895-442
2,6-Dichloro-4-pyridineboronic acid ≥97%
2,6-Dichloro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BCl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD09751352
CAS Number: 1072951-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR